Learn More
Statebourne Cryogenics™ Bulk Storage System Biosystem 12 Cryogenic Ics
Brand:  Statebourne Cryogenics™ 5801259
Additional Details : Weight : 0.80000kg
Product Code. 11497446
  • 60959.20 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
Manufactured from high quality stainless steel, Biosystem models are castor mounted with a lockable hinged lid. Large neck opening gives easy unrestricted access to samples. Storage of biological material can be carried out in either liquid phase at -196°C or under vapour phase conditions as low as -190°C. A choice from three base models in the range provides storage for up to 37,400 vials in one refrigerator. Safe, automated storage is provided with the security of having the system run and monitored by an integrated microprocessor based controller. Key pad touch control allows for easy set-up and continuous monitoring to provide complete security for your biological material. The Biosystem microprocessor based controller provides the user with the ultimate in sample security and container reliability. GLP traceability information is provided through accurate data logging facility. All inventory systems are supplied complete with plastic cryoboxes.All models have hot gas bypass option and daisy chain fill (saves LN2). Biosystem 12 and Biosystem 14 have vapour guard overfill protection and super chill (-190°C) option.
Dual solenoids fitted as standard to prevent LN2 overfill
  • As low as -190°C vapour phase storage temperature
  • Cryovial and straw inventory control systems, liquid or vapour
  • Custom built special storage systems available
  • CE marked in accordance with EC Medical Devices Directive
Bulk cryogenic inventory storage system.
Videos
Documents
View accessory products