Läs mer
Bumetrizole, TRC
Produktkod. 30337932
Change view
Kvantitet:
100 g
Produktkod. Kvantitet
30337932 100 g
1 options
Produktkod. 30337932 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB689680100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bumetrizole
CAS 3896-11-5
Molekylformel C17 H18 Cl N3 O
Kvantitet 100 g
Synonym p-Cresol, 2-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)- (7CI,8CI), 2-(5-Chloro-2H-benzotriazol-2-yl)-6-(1,1-dimethylethyl)-4-methylphenol, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chloro-2H-benzotriazole, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3'-tert-butyl-5'-methylphenyl)-5-chlorobenzotriazole, 2-(2'-Hydroxy-3'-tert-butyl-5'-methylphenyl)-5-chlorobenztriazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H-benzo-v-triazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chloro-2H-benzotriazole, 2-(3-tert-Butyl-2-hydroxy-5-methylphenyl)-5-chlorobenzotriazole, 2-(3-tert-Butyl-5-methyl-2-hydroxyphenyl)-5-chlorobenzotriazole, 2-(3'-tert-Butyl-2'-hydroxy-5'-methylphenyl)-5-chlorobenzotriazole, 2-(3'-tert-Butyl-5'-methyl-2'-hydroxyphenyl)-5-chlorobenzotriazole, 2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol, 2-(5-Chloro-2H-benzotriazol-2-yl)-6-tert-butyl-4-methylphenol, 2-[5-Chloro-2H-benzotriazol-2-yl]-4-methyl-6-(tert-butyl)phenol, 2-tert-Butyl-6-(5-chloro-2-benzotriazolyl)-4-methylphenol, 2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol, 5-Chloro-2-(2-hydroxy-3-tert-butyl-5-methylphenyl)-2H-benzotriazole, 5-Chloro-2-(3-tert-butyl-2-hydroxy-5-methylphenyl)benzotriazole, A-Light, ADK Stab LA 36, ADK Stab LA 36RG, Benazol PBKh, Bumetrizole, CBT, Cibatex LF, Eversorb 73, J 395, J 395 (light stabilizer), JC 30S, JF 79, Kemisorb 73, LA 36, Light Stabilizer 326, Lowilite 26, Mark LA 36, Seesorb 703, Songsorb 3260, Sumisorb 300, T 326, TNV 326, Tin 326, Tinogard AS, Tinuvin 326, Tomisorb 600, UV 326, Uvinul 3026, VS 550, Viosorb 550
InChI formel InChI=1S/C17H18ClN3O/c1-10-7-12(17(2,3)4)16(22)15(8-10)21-19-13-6-5-11(18)9-14(13)20-21/h5-9,22H,1-4H3
LEDER Cc1cc(c(O)c(c1)C(C)(C)C)n2nc3ccc(Cl)cc3n2
IUPAC-namn 2-tert-butyl-6-(5-chlorobenzotriazol-2-yl)-4-methylphenol
Molekylvikt (g/mol) 315.797
Formel vikt 315.1138