Läs mer
Buparlisib Hydrochloride, MedChemExpress
Produktkod. 16466656
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16466656 100 mg
16466348 10 mg
16438937 5 mg
16402467 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16466656 Leverantör Medchem Express Leverantörsnummer HY15180
MedChemExpress Buparlisib Hydrochloride (BKM120 Hydrochloride) is a pan-class I PI3K inhibitor, with IC50 of 52 nM/166 nM/116 nM/262 nM for p110α/p110β/p110δ/p110γ, respectively.
Buparlisib Hydrochloride (BKM120 Hydrochloride) is a pan-class I PI3K inhibitor, with IC50 of 52 nM/166 nM/116 nM/262 nM for p110α/p110β/p110δ/p110γ, respectively.
Specifikationer
Kemiskt namn eller material Buparlisib Hydrochloride
CAS 1312445-63-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C18H22ClF3N6O2
Kvantitet 100 mg
Synonym BKM120 HydrochlorideNVP-BKM120 Hydrochloride
Löslighetsinformation DMSO : ≥ 50 mg/mL (111.89 mM)
LEDER NC(N=C1)=CC(C(F)(F)F)=C1C2=NC(N3CCOCC3)=NC(N4CCOCC4)=C2.[H]Cl
Molekylvikt (g/mol) 446.85
Formel vikt 446.85
Procent renhet 96.98%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease