Läs mer
Thermo Scientific™ BupH™ Tris-HEPES-SDS Running Buffer
Handla allt Thermo Scientific ProdukterBeskrivning
Thermo Scientific™ BupH Tris Buffered Saline Packs are pouches of dry-blend powder that are each sufficient to make 500mL of Tris-NaCl buffer (TBS) for diluents and wash buffers in ELISA, Western blotting and general immunoassay procedures.
Highlights:
TBS— dissolved in 500mL of water, each pack makes 25mM Tris, 0.15M NaCl, pH 7.2
Convenient—dissolve contents of one envelope in water and the buffer is ready to use
Save time and trouble—no weighing, no pH adjustment, no need to stock individual components and no need to make and store large volumes of stock solutions in advance of daily needs
Long shelf life—stocking and storage of dry packs eliminates concerns about long-term stability of stock solutions
Eliminate variables—our quality control ensures that every pack will yield the same, consistent buffer
Recommended for:
Wash buffer and antibody diluent for ELISA, Western blotting and other immunoassays
Specifikationer
Specifikationer
| Kemiskt namn eller material | Running Buffer |
| Produktlinje | BupH |
| Kvantitet | 10 packs |