Läs mer
Buphenine Hydrochloride, LoGiCal
Specifikationer
Specifikationer
| Kemiskt namn eller material | Buphenine Hydrochloride |
| CAS | 849-55-8 |
| Analyt- eller komponentnamn | Buphenine Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C19 H25 N O2 . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Buphenine Hydrochloride, Nylidrin hydrochloride, Arlidin, Arlidin hydrochloride, Bufedon, Buphedrin, Buphenin hydrochloride, Buphenine hydrochl, 4-Hydroxy-alpha-[1-[(1-methyl-3-phenylpropyl)amino]ethyl]benzenemethanol hydrochloride, (+/-)-Nylidrin hydrochloride, Buphenine hydrochloride, Dilatal, Dilatol, Dilatol hydrochloride, Dilatropon, Dilydrin, NSC 142004, Nylidrine hydrochloride, Opino, Penitardon, Perdilatal, RHC 3432A, Rudilin, Rydrin, SKF 1700A, Suprifen Psb hydrochloride, Tocodilydrin, Tocodrin, Verina, p-Hydroxy-alpha-[1-[(1-methyl-3-phenylpropyl)amino]ethyl]benzyl alcohol hydrochloride |
| InChI formel | InChI=1S/C19H25NO2.ClH/c1-14(8-9-16-6-4-3-5-7-16)20-15(2)19(22)17-10-12-18(21)13-11-17;/h3-7,10-15,19-22H,8-9H2,1-2H3;1H |
| Lösningstyp | Neat |
| LEDER | Cl.CC(CCc1ccccc1)NC(C)C(O)c2ccc(O)cc2 |
| Visa mer |