Läs mer
Buphenine Hydrochloride, Mikromol™
Produktkod. 17951876
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
17951876 500 mg
1 options
Produktkod. 17951876 Leverantör Mikromol Leverantörsnummer MM0710.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Buphenine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Buphenine Hydrochloride
CAS 849-55-8
Analyt- eller komponentnamn Buphenine Hydrochloride
Frakt skick Room Temperature
Molekylformel C19 H25 N O2 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Buphenine Hydrochloride, Nylidrin hydrochloride, Arlidin, Arlidin hydrochloride, Bufedon, Buphedrin, Buphenin hydrochloride, Buphenine hydrochl, 4-Hydroxy-alpha-[1-[(1-methyl-3-phenylpropyl)amino]ethyl]benzenemethanol hydrochloride, (+/-)-Nylidrin hydrochloride, Buphenine hydrochloride, Dilatal, Dilatol, Dilatol hydrochloride, Dilatropon, Dilydrin, NSC 142004, Nylidrine hydrochloride, Opino, Penitardon, Perdilatal, RHC 3432A, Rudilin, Rydrin, SKF 1700A, Suprifen Psb hydrochloride, Tocodilydrin, Tocodrin, Verina, p-Hydroxy-alpha-[1-[(1-methyl-3-phenylpropyl)amino]ethyl]benzyl alcohol hydrochloride
InChI formel InChI=1S/C19H25NO2.ClH/c1-14(8-9-16-6-4-3-5-7-16)20-15(2)19(22)17-10-12-18(21)13-11-17;/h3-7,10-15,19-22H,8-9H2,1-2H3;1H
LEDER Cl.CC(CCc1ccccc1)NC(C)C(O)c2ccc(O)cc2
IUPAC-namn 4-[1-hydroxy-2-(4-phenylbutan-2-ylamino)propyl]phenol;hydrochloride
Molekylvikt (g/mol) 335.87
Kvantitet 500 mg
Formel vikt 335.1652 g/mol