Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Bupirimate |
| CAS | 41483-43-6 |
| CAS Max % | ≤ 100% |
| Beilstein | 758056 |
| Färg | Light Brown |
| Smältpunkt | 48°C to 52°C |
| Kokpunkt | 232°C |
| Synonym | 5-butyl-2-ethylamino-6-methylpyrimidin-4-yl dimethylsulfamate |
| Molekylvikt (g/mol) | 316.4 g/mol |
| Kvantitet | 100 mg |