Läs mer
Bupivacaine Hydrochloride, TRC
Produktkod. 30138608
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138608 5 g
30134576 500 mg
30009655 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB7805955G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18010-40-7
Molekylformel C18 H28 N2 O . Cl H
Molekylvikt (g/mol) 324.89
Synonym 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide monohydrochloride, 1-Butyl-2',6'-pipecoloxylidide hydrochloride, 1-Butyl-2',6'-pipecoloxylidide monohydrochloride, (±)-Bupivacaine hydrochloride, AH 2250, Bicain, Bupivacaine hydrochloride, LAC 43, Marcaina, Marcaine hydrochloride, Sensorcaine, NovaBupi
IUPAC-namn 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride
LEDER Cl.CCCCN1CCCCC1C(=O)Nc2c(C)cccc2C
Specifikationer
Kemiskt namn eller material Bupivacaine Hydrochloride
Kvantitet 5 g
InChI formel InChI=1S/C18H28N2O.ClH/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H
Formel vikt 324.1968