Läs mer
Bupivacaine Related Compound B, TRC
Produktkod. 30346266
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30346266 250 mg
1 options
Produktkod. 30346266 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB780600250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
CAS 65797-42-4
Molekylformel C14 H20 N2 O . Cl H
Kvantitet 250 mg
Synonym (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide Hydrochloride, Mepivacaine Hydrochloride Imp. B (EP) as Hydrochloride
InChI formel InChI=1S/C14H20N2O.ClH/c1-10-6-5-7-11(2)13(10)16-14(17)12-8-3-4-9-15-12;/h5-7,12,15H,3-4,8-9H2,1-2H3,(H,16,17);1H
LEDER Cl.Cc1cccc(C)c1NC(=O)C2CCCCN2
IUPAC-namn N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrochloride
Molekylvikt (g/mol) 268.78
Formel vikt 268.1342