Läs mer
Buserelin Acetate, TRC
Produktkod. 30006429
Change view
Kvantitet:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006429 1 mg
30006380 10 mg
30004307 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006429 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6897401MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68630-75-1
Synonym 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-1-9-Luteinizing hormone-releasing factor (swine) acetate (1:1)∣1-9-Luteinizing hormone-releasing factor (swine), 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-, monoacetate (salt) (9CI)∣Luteinizing hormone-releasing factor (pig), 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-10-deglycinamide-, monoacetate (salt)∣1-9-Luteinizing hormone-releasing factor (pig), 6-[O-(1,1-dimethylethyl)-D-serine]-9-(N-ethyl-L-prolinamide)-, monoacetate (salt)∣Buserelin acetate∣Conceptal∣Estomal∣Gonaxal∣Sprecur MP∣Suprafact∣Suprecur∣Supremon
Specifikationer
Formatera Neat
Molekylformel C62H90N16O15
MDL-nummer MFCD01716229
Molekylvikt (g/mol) 1299.48, 1299.50
InChI-nyckel PYMDEDHDQYLBRT-DRIHCAFSSA-N
LEDER CC(O)=O.CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCN=C(N)N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](COC(C)(C)C)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)[C@H](CC1=CN=CN1)NC(=O)[C@@H]1CCC(=O)N1
IUPAC-namn (2S)-1-[(2S)-2-[(2S)-2-[(2R)-3-(tert-butoxy)-2-[(2S)-2-[(2S)-3-hydroxy-2-[(2S)-2-[(2S)-3-(1H-imidazol-5-yl)-2-{[(2S)-5-oxopyrrolidin-2-yl]formamido}propanamido]-3-(1H-indol-3-yl)propanamido]propanamido]-3-(4-hydroxyphenyl)propanamido]propanamido]-4-methylpentanamido]-5-[(diaminomethylidene)amino]pentanoyl]-N-ethylpyrrolidine-2-carboxamide; acetic acid