Läs mer
Busulfan, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Busulfan |
| CAS | 55-98-1 |
| Molekylformel | C6 H14 O6 S2 |
| Kvantitet | 100 mg |
| Synonym | Busulfan, Butane-1,4-diyl di(methanesulfonate), 1,4-Butanediol, 1,4-dimethanesulfonate (ACI), 1,4-Butanediol, dimethanesulfonate (8CI, 9CI), Methanesulfonic acid, tetramethylene ester (6CI), 1,4-Bis(methanesulfonoxy)butane, 1,4-Bis(methanesulfonyloxy)butane, 1,4-Butanediol dimesulfonate, 1,4-Butanediol dimesylate, 1,4-Dibutanediol dimethanesulfonate, 1,4-Dimethanesulfonoxybutane, 1,4-Dimethylsulfonyloxybutane, 2041CB, 4-Methylsulfonyloxybutyl methanesulfonate, AN 33501, Busilvex, Busulfex, Busulphan, Butane-1,4-diyl bis(methanesulfonate), CB 2041, Citosulfan, Glyzophrol, GT 2041, GT 41, Leucosulfan, Mablin, MeSH ID: D002066, Mielevcin, Mielosan, Mielucin, Milecitan, Mileran, Misulban, Mitostan, Myeloleukon, Myelosan, Mylecytan, Myleran, Myran, NCI C01592, NSC 750, Sulfabutin, Sulphabutin, Tetramethylene bis[methanesulfonate], X 149 |
| InChI formel | InChI=1S/C6H14O6S2/c1-13(7,8)11-5-3-4-6-12-14(2,9)10/h3-6H2,1-2H3 |
| LEDER | CS(=O)(=O)OCCCCOS(=O)(=O)C |
| IUPAC-namn | 4-methylsulfonyloxybutyl methanesulfonate |
| Molekylvikt (g/mol) | 246.3 |
| Formel vikt | 246.0232 |