Läs mer
Butacaine Sulphate, TRC
Produktkod. 30345162
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30345162 1 g
1 options
Produktkod. 30345162 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6900001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butacaine Sulfate
CAS 149-15-5
Molekylformel 2 C18 H30 N2 O2 . H2 O4 S
Kvantitet 1 g
Synonym 1-Propanol, 3-(dibutylamino)-, 1-(4-aminobenzoate), sulfate (2:1), 1-Propanol, 3-(dibutylamino)-, 4-aminobenzoate (ester), sulfate (2:1) (salt) (9CI), 1-Propanol, 3-(dibutylamino)-, p-aminobenzoate (ester), sulfate (2:1) (8CI), Butacaine sulfate (6CI), 3-(p-Aminobenzoxy)-1-di-n-butylaminopropane sulfate, 3'-Dibutylaminopropyl 4-aminobenzoate sulfate, Butacaine hemisulfate, Butyn Sulfate, Dibutylaminopropyl p-aminobenzoate sulfate
InChI formel InChI=1S/2C18H30N2O2.H2O4S/c2*1-3-5-12-20(13-6-4-2)14-7-15-22-18(21)16-8-10-17(19)11-9-16;1-5(2,3)4/h2*8-11H,3-7,12-15,19H2,1-2H3;(H2,1,2,3,4)
LEDER CCCCN(CCCC)CCCOC(=O)c1ccc(N)cc1.CCCCN(CCCC)CCCOC(=O)c2ccc(N)cc2.OS(=O)(=O)O
IUPAC-namn 3-(dibutylamino)propyl 4-aminobenzoate;sulfuric acid
Molekylvikt (g/mol) 710.96
Formel vikt 710.4288