Learn More
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Catalog Number p-3777725
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
77-79-2
C4H6O2S
118.16
MFCD00005481
MBDNRNMVTZADMQ-UHFFFAOYSA-N
3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569
6498
2,5-dihydrothiophene 1,1-dioxide
C1C=CCS1(=O)=O
77-79-2
C4H6O2S
118.16
MFCD00005481
MBDNRNMVTZADMQ-UHFFFAOYSA-N
3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569
6498
2,5-dihydrothiophene 1,1-dioxide
C1C=CCS1(=O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 63.0°C to 66.0°C
Color Brown or Orange to Yellow
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 17, V,1, 177
Fieser 02,389; 03,272; 04,468; 12,455
Merck Index 15,9086
Solubility Information Solubility in water: 130g/L (20°C). Other solubilities: soluble in ethanol, acetone, chloroform, benzene
Formula Weight 118.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Butadiene sulfone
Show More Show Less
compliance-icons
Product Identifier
  • Butadiene sulfone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.