Läs mer
Butalamine Hydrochloride, LoGiCal
Produktkod. 30461382
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461382 10 mg
1 options
Produktkod. 30461382 Leverantör LoGiCal Leverantörsnummer LGCFOR1845.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butalamine Hydrochloride
CAS 56974-46-0
Analyt- eller komponentnamn Butalamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C18 H28 N4 O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Butalamine Hydrochloride, 1,2-Ethanediamine, N1,N1-dibutyl-N2-(3-phenyl-1,2,4-oxadiazol-5-yl)-, hydrochloride (1:1), 1,2-Ethanediamine, N,N-dibutyl-N'-(3-phenyl-1,2,4-oxadiazol-5-yl)-, monohydrochloride, Adrevil, Adrevil forte, Butalamine hydrochloride, Hemotrope, LA 1221, Surem, Surheme
InChI formel InChI=1S/C18H28N4O.ClH/c1-3-5-13-22(14-6-4-2)15-12-19-18-20-17(21-23-18)16-10-8-7-9-11-16;/h7-11H,3-6,12-15H2,1-2H3,(H,19,20,21);1H
Lösningstyp Neat
LEDER Cl.CCCCN(CCCC)CCNc1onc(n1)c2ccccc2
IUPAC-namn N',N'-dibutyl-N-(3-phenyl-1,2,4-oxadiazol-5-yl)ethane-1,2-diamine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 352.9
Kvantitet 10 mg
Formel vikt 352.203 g/mol