Läs mer
Butamirate, TRC
Produktkod. 30343362
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30343362 25 mg
1 options
Produktkod. 30343362 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB69986025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butamirate
CAS 18109-80-3
Molekylformel C18H29NO3
Kvantitet 25 mg
Synonym Butamyrate, 2-[2-(Diethylamino)ethoxy]ethyl 2-Phenylbutyrate, 2-[2-(Diethylamino)ethoxy]-ethanol 2-Phenylbutyrate, 2-Phenyl-butyric Acid 2-[2-(Diethylamino)ethoxy]ethyl Ester, alpha-Ethyl-benzeneacetic Acid
InChI formel InChI=1S/C18H29NO3/c1-4-17(16-10-8-7-9-11-16)18(20)22-15-14-21-13-12-19(5-2)6-3/h7-11,17H,4-6,12-15H2,1-3H3
LEDER CCC(C(=O)OCCOCCN(CC)CC)c1ccccc1
IUPAC-namn 2-[2-(diethylamino)ethoxy]ethyl 2-phenylbutanoate
Molekylvikt (g/mol) 307.43
Formel vikt 307.2147