Läs mer
Butanilicaine Hydrochloride, TRC
Produktkod. 30343660
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30343660 100 mg
1 options
Produktkod. 30343660 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB693055100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butanilicaine Hydrochloride
CAS 6027-28-7
Molekylformel C13 H19 Cl N2 O . Cl H
Kvantitet 100 mg
Synonym 1-(Butylaminoacetylamino)-2-chloro-6-methylbenzene Hydrochloride, 2-(Butylamino)-6'-chloro-o-acetoluidide, 2-Butylamino-6'-chloro-o-acetotoluidide Hydrochloride, ω-n-Butylaminoacetic acid 2-methyl-6-chloroanilide Hydrochloride
InChI formel InChI=1S/C13H19ClN2O.ClH/c1-3-4-8-15-9-12(17)16-13-10(2)6-5-7-11(13)14;/h5-7,15H,3-4,8-9H2,1-2H3,(H,16,17);1H
LEDER Cl.CCCCNCC(=O)Nc1c(C)cccc1Cl
IUPAC-namn 2-(butylamino)-N-(2-chloro-6-methyl-phenyl)acetamide;hydrochloride
Molekylvikt (g/mol) 291.22
Formel vikt 290.0953