Läs mer
Butaperazine Dihydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Butaperazine Dihydrochloride |
| CAS | 80463-18-9 |
| Molekylformel | C24H31N3OS .2HCl |
| Kvantitet | 100 mg |
| Synonym | 1-[10-[3-(4-Methyl-1-piperazinyl)propyl]-10H-phenothiazin-2-yl]-1-butanone Dihydrochloride, 2-Butyryl-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine Dihydrochloride, AHR 3000 Dihydrochloride, AHR 712 Dihydrochloride, Bayer 1362 Dihydrochloride, Butyrylperazine Dihydrochloride, Emerex Dihydrochloride, Megalectil Dihydrochloride, (1-oxobutyl)-perazine Dihydrochloride: Randolectil Dihydrochloride, Repoise Dihydrochloride, Riker 595 Dihydrochloride, Tyrylen Dihydrochloride |
| InChI formel | InChI=1S/C24H31N3OS/c1-3-7-22(28)19-10-11-24-21(18-19)27(20-8-4-5-9-23(20)29-24)13-6-12-26-16-14-25(2)15-17-26/h4-5,8-11,18H,3,6-7,12-17H2,1-2H3 |
| LEDER | O=C(CCC)C1=CC=C(SC(C=CC=C2)=C2N3CCCN4CCN(C)CC4)C3=C1 |
| IUPAC-namn | 1-[10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazin-2-yl]butan-1-one |
| Molekylvikt (g/mol) | 409.5923646 |