Läs mer
Buthiobate, TRC
Produktkod. 30334002
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30334002 10 mg
1 options
Produktkod. 30334002 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB99958010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Buthiobate
CAS 51308-54-4
Molekylformel C21 H28 N2 S2
Kvantitet 10 mg
Synonym Dimethylethyl)Phenyl]Methyl Ester, Butiobate, Den, Denmert, N-(3-Pyridyl) S-Butyl S'-(P-Tert-Butylbenzyl)Imidodithiocarbonate, S 1358, (E)-Butyl 4-(Tert-Butyl)Benzyl Pyridin-3-Ylcarbonimidodithioate
InChI formel InChI=1S/C21H28N2S2/c1-5-6-14-24-20(23-19-8-7-13-22-15-19)25-16-17-9-11-18(12-10-17)21(2,3)4/h7-13,15H,5-6,14,16H2,1-4H3
LEDER Ccccsc(=Nc1Cccnc1)Scc2Ccc(Cc2)C(C)(C)C
IUPAC-namn 1-[(4-tert-butylphenyl)methylsulfanyl]-1-butylsulfanyl-N-pyridin-3-ylmethanimine
Molekylvikt (g/mol) 372.59
Formel vikt 372.1694