Läs mer
Butropium Bromide, TRC
Produktkod. 30004870
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30004870 5 mg -
30007163 2.5 mg 2.5mg
30003544 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004870 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6900805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29025-14-7
Molekylformel C28 H38 N O4 . Br
Molekylvikt (g/mol) 532.51
Synonym 8-Azoniabicyclo[3.2.1]octane, 8-[(4-butoxyphenyl)methyl]-3-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-8-methyl-, bromide (1:1), (3-endo)-, 1αH,5αH-Tropanium, 8-(p-butoxybenzyl)-3α-hydroxy-, bromide, (-)-tropate (8CI), 8-Azoniabicyclo[3.2.1]octane, 8-[(4-butoxyphenyl)methyl]-3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-, bromide, [3(S)-endo]- (9CI), (p-Butoxybenzyl)hyoscyaminium bromide, BHB, Butropium bromide, Coliopan, N-(Butoxybenzyl)hyoscyamine bromide
IUPAC-namn [(1R,5S)-8-[(4-butoxyphenyl)methyl]-8-methyl-8-azoniabicyclo[3.2.1]octan-3-yl] (2S)-3-hydroxy-2-phenylpropanoate;bromide
LEDER [Br-].CCCCOc1ccc(C[N+]2(C)[C@@H]3CC[C@H]2C[C@H](C3)OC(=O)[C@H](CO)c4ccccc4)cc1
Specifikationer
Kemiskt namn eller material Butropium Bromide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C28H38NO4.BrH/c1-3-4-16-32-25-14-10-21(11-15-25)19-29(2)23-12-13-24(29)18-26(17-23)33-28(31)27(20-30)22-8-6-5-7-9-22;/h5-11,14-15,23-24,26-27,30H,3-4,12-13,16-20H2,1-2H3;1H/q+1;/p-1/t23-,24+,26+,27-,29?;/m1./s1
Formel vikt 531.1984
Procent renhet >95%