Läs mer
Butyl 3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl Carbonate, TRC
Produktkod. 30417152
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417152 250 mg
30408943 25 mg
30415552 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417152 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS240640250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1305319-70-3
Molekylformel C20H22Cl2O5
Molekylvikt (g/mol) 413.29
Synonym Spirobudiclofen, Carbonic acid, butyl 3-(2,4-dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl ester, Butyl 3-(2,4-dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl carbonate, Carbonic acid butyl 3-(2,4-dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl ester, Spirobudifen
IUPAC-namn butyl [3-(2,4-dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl] carbonate
LEDER CCCCOC(=O)OC1=C(C(=O)OC12CCCCC2)c3ccc(Cl)cc3Cl
Specifikationer
Kemiskt namn eller material Butyl 3-(2,4-Dichlorophenyl)-2-oxo-1-oxaspiro[4.5]dec-3-en-4-yl Carbonate
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C20H22Cl2O5/c1-2-3-11-25-19(24)26-17-16(14-8-7-13(21)12-15(14)22)18(23)27-20(17)9-5-4-6-10-20/h7-8,12H,2-6,9-11H2,1H3
Formel vikt 412.0844
Procent renhet >95%