Läs mer
Butyl Paraben-13C6, TRC
Produktkod. 30009958
Change view
Kvantitet:
0.5 mg
1 mg
2.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009958 0.5 mg
30008782 2.5 mg
30004285 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009958 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6936030.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1416711-53-9
Molekylformel 13C6 C5 H14 O3
Molekylvikt (g/mol) 200.18
Synonym 4-Hydroxybenzoic acid-n-butyl ester 13C6 (ring 13C6), Benzoic-1,2,3,4,5,6-13C6 acid, 4-hydroxy-, butyl ester (ACI), Butyl 4-hydroxybenzoate-1,2,3,4,5,6-13C6 (ACI), DTXSID 90894091
IUPAC-namn butyl 4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-carboxylate
LEDER CCCCOC(=O)[13c]1[13cH][13cH][13c](O)[13cH][13cH]1
Specifikationer
Kemiskt namn eller material n-Butyl paraben ring-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 0.5 mg
InChI formel InChI=1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3/i4+1,5+1,6+1,7+1,9+1,10+1
Formel vikt 200.1144
Procent renhet >95%