Learn More
Wheaton™ Rubber Freeze Dry Stoppers
Product Code. 13550970 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
14 mm
20 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
13550970 14 mm Pack of 1000
14833592 20 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13550970 Supplier Wheaton™ Supplier No. 600005
  • ONLINE EXCLUSIVE PRICE
    1865.00 SEK
    1463.72 SEK / Paket med 1000
    Save 401.28 SEK
    21% Av
I lager
Add to basket
Add to basket
Provides for safe and secure closure of bottles and vials. Wheaton™ Rubber Freeze Dry Stoppers are manufactured from butyl rubber.
  • Available in two sizes
  • Freeze dry stoppers are push-in (not screw thread)
Specifications
Diameter (Metric) 14 mm
For Use With (Equipment) Diagnostic vials
Color Gray
Material Butyl Rubber
Product Type Plug Stopper