Läs mer
Butyl sec-Butyl Phthalate, TRC
Produktkod. 30335091
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30335091 100 mg
1 options
Produktkod. 30335091 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB692440100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butyl sec-Butyl Phthalate
CAS 97621-09-5
Molekylformel C16H22O4
Kvantitet 100 mg
Synonym 1,2-Benzenedicarboxylic Acid 1-Butyl 2-(1-methylpropyl) Ester, Phthalic Acid Butyl sec-Butyl Ester
InChI formel InChI=1S/C16H22O4/c1-4-6-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12(3)5-2/h7-10,12H,4-6,11H2,1-3H3
LEDER CCCCOC(=O)c1ccccc1C(=O)OC(C)CC
IUPAC-namn 2-O-butan-2-yl 1-O-butyl benzene-1,2-dicarboxylate
Molekylvikt (g/mol) 278.34
Formel vikt 278.1518