Läs mer
Butyltrimethylammonium Chloride, TRC
Produktkod. 30331493
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30331493 5 g
1 options
Produktkod. 30331493 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6937805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butyltrimethylammonium Chloride
CAS 14251-72-0
Molekylformel C7H18ClN
Kvantitet 5 g
Synonym N-Butyl-N,N,N-trimethylammonium Chloride, N,N,N-Trimethyl-1-butanaminium Chloride
InChI formel InChI=1S/C7H18N.ClH/c1-5-6-7-8(2,3)4;/h5-7H2,1-4H3;1H/q+1;/p-1
LEDER [Cl-].CCCC[N+](C)(C)C
IUPAC-namn butyl(trimethyl)azanium;chloride
Molekylvikt (g/mol) 151.68
Formel vikt 151.1128