Learn More
Butyraldehyde, Sigma-Aldrich™
Product Code. 15886290
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886290 100 mL 100mL
15818590 1 L 1 L
15828590 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15886290 Supplier Sigma Aldrich Supplier No. 8015550100
 Restricted Item
Beräknad leverans: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-72-8
Specifications
Melting Point -99°C
Density 0.80 g/cm3 at 20°C
Boiling Point 75°C to 76°C/1013 hPa
MDL Number MFCD00007023
Quantity 100 mL
Synonym Butyraldehyde, Butanal, Butyraldehyde
Molecular Weight (g/mol) 72.11
Grade Research