Läs mer
Butyramide, TRC
Produktkod. 30337911
Change view
Kvantitet:
250 g
Produktkod. Kvantitet
30337911 250 g
1 options
Produktkod. 30337911 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB812313250G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Butyramide
CAS 541-35-5
Molekylformel C4H9NO
Kvantitet 250 g
Synonym Butanimidic Acid, NSC 8424, n-Butylamide, n-Butyramide
InChI formel InChI=1S/C4H9NO/c1-2-3-4(5)6/h2-3H2,1H3,(H2,5,6)
LEDER CCCC(=O)N
IUPAC-namn butanamide
Molekylvikt (g/mol) 87.12
Formel vikt 87.0684