Läs mer
Butyrolactone 3, TRC
Produktkod. 30009303
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009303 1 mg
30011480 50 mg
30008464 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009303 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB7610101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 778649-18-6
Molekylformel C9 H12 O4
Molekylvikt (g/mol) 184.189
Synonym (2R,3S)-rel-Tetrahydro-4-methylene-5-oxo-2-propyl-3-furancarboxylic Acid, 4-Methylene-5-oxo-2-propyl-tetrahydrofuran-3-carboxylic Acid, 3-Furancarboxylic acid, tetrahydro-4-methylene-5-oxo-2-propyl-, (2R,3S)-rel-, rel-(2R,3S)-Tetrahydro-4-methylene-5-oxo-2-propyl-3-furancarboxylic acid, 4-Methylene-5-oxo-2-propyl-tetrahydrofuran-3-carboxylic acid, Butyrolactone 3, MB 3
IUPAC-namn (2S,3R)-4-methylene-5-oxo-2-propyl-tetrahydrofuran-3-carboxylic acid
LEDER CCC[C@@H]1OC(=O)C(=C)[C@H]1C(=O)O
Specifikationer
Kemiskt namn eller material Butyrolactone 3
Kvantitet 1 mg
InChI formel InChI=1S/C9H12O4/c1-3-4-6-7(8(10)11)5(2)9(12)13-6/h6-7H,2-4H2,1H3,(H,10,11)/t6-,7+/m0/s1
Formel vikt 184.0736