Learn More
Jaytec Glass™ Butyrometers Tube
Brand: Jaytec Glass™ MP.676
Additional Details : Weight : 0.01000kg
Description
Manufactured to BS 696.For testing fat content in dairy products, using the Gerber method.
All products are for research use only unless otherwise indicated.
Specifications
10.8 mL |