Learn More
BW B70C, TRC
Product Code. 30337187
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30337187 100 mg 100mg
1 options
Product Code. 30337187 Supplier Toronto Research Chemicals Supplier No. TRCB820010100MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
CAS 134470-38-5
Molecular Formula C17 H17 F N2 O3
Quantity 100 mg
Synonym BW B70C, N-[3-[3-(4-Fluorophenoxy)phenyl]-1-methyl-2-propen-1-yl]-N-hydroxyurea, BW-B 70C, N-[3-[3-(4-fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxy-urea
InChI Formula InChI=1S/C17H17FN2O3/c1-12(20(22)17(19)21)5-6-13-3-2-4-16(11-13)23-15-9-7-14(18)8-10-15/h2-12,22H,1H3,(H2,19,21)/b6-5+
SMILES CC(\C=C\c1cccc(Oc2ccc(F)cc2)c1)N(O)C(=O)N
IUPAC Name 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
Molecular Weight (g/mol) 316.327
Formula Weight 316.122