missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ BX 430
GREENER_CHOICE

Produktkod. 18793178
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18793178 10 mg
18759708 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18793178 Leverantör Tocris Bioscience™ Leverantörsnummer 5545/10

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Selective P2X4 allosteric antagonist

BX 430 is a selective P2X4 allosteric antagonist (IC50 = 0.54 μM). Exhibits >10-fold selectivity for human P2X4 receptors over P2X1, P2X2, P2X3, P2X5 and P2X7 receptors. Binds noncompetitively at extracellular allosteric site. Effective at human and zebrafish, but not mouse and rat P2X4 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-[2,6-Dibromo-4-(1-methylethyl)phenyl]-N′-(3-pyridinyl)urea
CAS 688309-70-8
Kvantitet 10 mg
Mål P2X Receptor Antagonists
Molekylformel C15H15Br2N3O
Formel vikt 413.11
Renhet >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.