missing translation for 'onlineSavingsMsg'
Läs mer

BX471 hydrochloride, MedChemExpress

Produktkod. 16441227
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16441227 50 mg
16474978 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16441227 Leverantör Medchem Express Leverantörsnummer HY12080A

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.

BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.

Specifikationer

Kemiskt namn eller material BX471 hydrochloride
CAS 288262-96-4
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C21H25Cl2FN4O3
Kvantitet 50 mg
Synonym ZK-811752 hydrochloride
Löslighetsinformation DMSO : 150 mg/mL (318.23 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
LEDER O=C(N)NC1=CC(Cl)=CC=C1OCC(N2[C@H](C)CN(CC3=CC=C(F)C=C3)CC2)=O.[H]Cl
Molekylvikt (g/mol) 471.35
Formel vikt 471.35
Procent renhet 99.51%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.