Läs mer
BX517, MedChemExpress
Produktkod. 16478527
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16478527 5 mg 5mg
16445888 10 mg -
16442037 50 mg 50mg
16413396 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16478527 Supplier Medchem Express Supplier No. HY138425mg
MedChemExpress BX517 is a potent and selective inhibitor of PDK1 with IC50 of 6 nM.
BX517 is a potent and selective inhibitor of PDK1 with IC50 of 6 nM.
Specifications
Kemiskt namn eller material BX517
Fysisk form Solid
CAS 850717-64-5
Färg Deep Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C15H14N4O2
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : ≥ 27 mg/mL (95.64 mM)
LEDER NC(NC1=CC=C(NC(/C2=C(C)\C3=CC=CN3)=O)C2=C1)=O
Molekylvikt (g/mol) 282.3
Formel vikt 282.3
Procent renhet 98.0%