Läs mer
(+)-Byakangelicin, TRC
Produktkod. 30344540
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30344540 50 mg
1 options
Produktkod. 30344540 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB81092850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Byakangelicin
CAS 482-25-7
Molekylformel C17 H18 O7
Kvantitet 50 mg
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, 9-[(2R)-2,3-dihydroxy-3-methylbutoxy]-4-methoxy-, 7H-Furo[3,2-g][1]benzopyran-7-one, 9-(2,3-dihydroxy-3-methylbutoxy)-4-methoxy-, (R)- (8CI), Biacangelicin (7CI), (+)-Byakangelicin, 5-Methoxy-8-(2,3-dihydroxy-3-methylbutoxy)psoralene, 9-[(2R)-2,3-Dihydroxy-3-methylbutoxy]-4-methoxy-7H-furo[3,2-g][1]benzopyran-7-one, Bjacangelicin, Bjakangelicin, Byakangelicin, Byankagelicine
InChI formel InChI=1S/C17H18O7/c1-17(2,20)11(18)8-23-16-14-10(6-7-22-14)13(21-3)9-4-5-12(19)24-15(9)16/h4-7,11,18,20H,8H2,1-3H3/t11-/m1/s1
LEDER COc1c2C=CC(=O)Oc2c(OC[C@@H](O)C(C)(C)O)c3occc13
IUPAC-namn 9-[(2R)-2,3-dihydroxy-3-methylbutoxy]-4-methoxyfuro[3,2-g]chromen-7-one
Molekylvikt (g/mol) 334.32
Formel vikt 334.1053