Learn More
Grant Bio™ Bypass Unit
Bypass to overcome flow restrictions
Brand:  Grant Bio™ RC BYP
Additional Details : Weight : 0.50000kg
Product Code. 11416517
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Bypass to overcome flow restrictions (flow <1L/min), e.g., in narrow tubes or small cells for Grant RC series high precision recirculating chillers.
Specifications
Specifications
< 1L/min