Läs mer
C-7280948, MedChemExpress
Produktkod. 16412967
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16412967 50 mg
16406908 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16412967 Leverantör Medchem Express Leverantörsnummer HY15890
MedChemExpress C-7280948 is a selective and potent protein methyltransferase1 (PRMT1) inhibitor with an IC50 value of 12.75 μM.
C-7280948 is a selective and potent protein methyltransferase1 (PRMT1) inhibitor with an IC50 value of 12.75 μM.
Specifikationer
Kemiskt namn eller material C-7280948
CAS 587850-67-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C14H16N2O2S
Kvantitet 50 mg
Löslighetsinformation DMSO : ≥ 38 mg/mL (137.51 mM)
LEDER O=S(C1=CC=C(N)C=C1)(NCCC2=CC=CC=C2)=O
Molekylvikt (g/mol) 276.35
Formel vikt 276.35
Procent renhet 98.31%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death