Läs mer
Tocris Bioscience™ C 75
GREENER_CHOICE
Produktkod. 18065672
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18065672 10 mg
18075172 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18065672 Leverantör Tocris Bioscience™ Leverantörsnummer 2489/10
Potent fatty acid synthase inhibitor; proapoptotic
Synthetic inhibitor of fatty acid synthase (FASN); inhibits fatty acid synthesis in vitro and in vivo. Displays anorectic effects. Induces apoptosis in MCF-7 xenografts and exhibits anti-tumor activity.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 191282-48-1
Molekylformel C14H22O4
Specifikationer
Kemiskt namn eller material C 75
Kvantitet 10 mg
Formel vikt 254.32
Renhet >99%
Molekylvikt (g/mol) 254.33