Läs mer
Tocris Bioscience™ C 87
TNF-α inhibitor
Brand: Tocris Bioscience™ 5484/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
C 87 is a TNF-α inhibitor (Kd = 110 nM for hTNF-α). Inhibits TNF-α-induced cytotoxicity in L929 fibroblast cells. Attenuates inflammation and increases survival in a mouse hepatitis model.Specifikationer
| C 87 | |
| 3-Phenyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(4-chloro-3-nitrophenyl)hydrazone] | |
| Soluble to 20mM in DMSO | |
| Bioactive Small Molecule |
| 332420-90-3 | |
| 50 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use