Läs mer
C-Desmethyl Atenolol, TRC
Produktkod. 30360110
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360110 100 mg
30356525 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360110 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD290800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797116-92-7
Molekylformel C13 H20 N2 O3
Molekylvikt (g/mol) 252.31
Synonym 2-[4-[(2RS)-3-(Ethylamino)-2-hydroxypropoxy]phenyl]acetamide, Atenolol Imp. I (EP), 2-{4-[3-(Ethylamino)-2-hydroxypropoxy]phenyl}acetamide, Benzeneacetamide, 4-[3-(ethylamino)-2-hydroxypropoxy]- (ACI), 4-[3-(Ethylamino)-2-hydroxypropoxy]benzeneacetamide (ACI)
IUPAC-namn 2-[4-[3-(ethylamino)-2-hydroxypropoxy]phenyl]acetamide
LEDER CCNCC(O)COc1ccc(CC(=O)N)cc1
Specifikationer
Kemiskt namn eller material C-Desmethyl Atenolol
Kvantitet 100 mg
InChI formel InChI=1S/C13H20N2O3/c1-2-15-8-11(16)9-18-12-5-3-10(4-6-12)7-13(14)17/h3-6,11,15-16H,2,7-9H2,1H3,(H2,14,17)
Formel vikt 252.1474