Läs mer
Eurofins DiscoverX c-Kit (V560G) Protein, Active
Beskrivning
Binding of stem cell factor (SCF) to C-Kit receptor tyrosine kinase induces homodimerization and intermolecular tyrosine phosphorylation of the receptor, creating docking sites for a number of SH2-containing signal transduction molecules. These include phosphatidylinositol 3'-kinase (PI 3K) and phospholipase C-1, Stat1, the Src family kinases Src, Lyn, and Fyn, and the GTPase activating protein GAP. Mutation in Kit which result in ligand-independent activation of the kinase were found to be associated with human gastrointestinal stromal tumors and mastocytosis.
GenBank X06182
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 76.6 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Uttryckssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Proteinform | Truncated |
| Renhet eller kvalitetsklass | ≥71% |
| Protein | Kit (V560G), active |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.