Läs mer
C10-C16 Alkyl Ethoxylate Sulfuric Acid Sodium Salt, TRC
Produktkod. 30428195
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30428195 10 g
1 options
Produktkod. 30428195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS22472010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material C10-C16 Alkyl Ethoxylate Sulfuric Acid Sodium Salt
CAS 68585-34-2
Kvantitet 10 g
Synonym Alcohol Ethoxysulfate (Sodium Salt)