missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
C12orf4 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21423JF646
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
C12orf4 Polyclonal antibody specifically detects C12orf4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifikationer
| C12orf4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| chromosome 12 open reading frame 4, hypothetical protein LOC57102 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C12orf4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57102 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction