missing translation for 'onlineSavingsMsg'
Learn More

C13orf30 Rabbit anti-Canine, Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25583-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884135

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C13orf30
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N7L0
C13orf30, FAM216B
Rabbit
IgG
20 μL
-20°C
Primary
100682941, 144809
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Unconjugated
FAM216B
Liquid
FAM216B
C13orf30 Fusion Protein Ag22353
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Canine, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers