missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C13orf30 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C13orf30 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884135
|
Proteintech
25583-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874135
|
Proteintech
25583-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
C13orf30 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Canine, Human | |
Q8N7L0 | |
100682941, 144809 | |
C13orf30 Fusion Protein Ag22353 | |
Primary | |
-20°C | |
FAM216B |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C13orf30, FAM216B | |
FAM216B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |