missing translation for 'onlineSavingsMsg'
Learn More

C14orf138 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24722-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883205

  • 1355.00 SEK / 20µL
Estimated Shipment: 22-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

C14orf138, also named as VCPKMT or METTL21D, is 229 amino acid protein, which belongs to the methyltransferase superfamily. C14orf138 localizes in the cytoplasm and interacts with ALKBH6. C14orf138 as a Protein-lysine N-methyltransferase that specifically trimethylates ′Lys-315′ of VCP/p97; this modification may decrease VCP ATPase activity. C14orf138 exists as five isoforms. The calculated molecular weight of C14orf138 is 26 kDa, but we always detected a 46-50 kDa protein by western blot.
Specifications

Specifications

C14orf138
Polyclonal
Unconjugated
VCPKMT
C14orf138
Rabbit
Antigen Affinity Chromatography
RUO
79609
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H867
VCPKMT
C14orf138 Fusion Protein Ag20154
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers