missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20801-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
C16orf13 | |
Polyclonal | |
Unconjugated | |
METTL26 | |
C16orf13, JFP2, UPF0585 protein C16orf13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
302998, 68347, 84326 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q497C3, Q96S19, Q9DCS2 | |
Mettl26 | |
C16orf13 Fusion Protein Ag14718 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |