missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf93 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C16orf93 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883945
|
Proteintech
25364-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16873945
|
Proteintech
25364-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C16orf93, also named as CCDC189, is a coiled-coil domain-containing protein which has four isoforms with MW 38 kDa, 34 kDa, 12 kDa and 20 kDa.Specifications
C16orf93 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
A1A4V9 | |
90835 | |
C16orf93 Fusion Protein Ag21676 | |
Primary | |
-20°C | |
CCDC189 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C16orf93, CCDC189 | |
CCDC189 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |