missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf81 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10162-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.Specifications
C17orf81 | |
Polyclonal | |
Unconjugated | |
ELP5 | |
C17orf81, DERP6, ELP5, HSPC002, MST071, MSTP071, S phase 2 protein, S-phase 2 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23587 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TE02 | |
ELP5 | |
C17orf81 Fusion Protein Ag0194 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |