missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf81 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C17orf81 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826363
|
Proteintech
10162-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816363
|
Proteintech
10162-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.Specifications
C17orf81 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q8TE02 | |
23587 | |
C17orf81 Fusion Protein Ag0194 | |
Primary | |
-20°C | |
ELP5 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C17orf81, DERP6, ELP5, HSPC002, MST071, MSTP071, S phase 2 protein, S-phase 2 protein | |
ELP5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |