missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27382-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
C19orf12 | |
Polyclonal | |
Unconjugated | |
C19orf12 | |
C19orf12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83636 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NSK7 | |
C19orf12 | |
C19orf12 Fusion Protein Ag26555 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |