missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25514-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to C19orf70.
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane.Specifications
C19orf70 | |
Polyclonal | |
Unconjugated | |
MICOS13 | |
C19orf70, MIC13, P117, Protein P117, Protein QIL1, QIL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
125988, 224904 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5XKP0 | |
Micos13 | |
C19orf70 Fusion Protein Ag22003 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |